missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ACTL7A Full-length ORF (NP_006678.1, 1 a.a. - 435 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00010881-P01.25ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16165792

  • 528.00€ / 25 microgramos
Fecha estimada de envío: 03-11-2023
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

The protein encoded by this gene is a member of a family of actin-related proteins (ARPs) which share significant amino acid sequence identity to conventional actins. Both actins and ARPs have an actin fold, which is an ATP-binding cleft, as a common feature. The ARPs are involved in diverse cellular processes, including vesicular transport, spindle orientation, nuclear migration and chromatin remodeling. This gene (ACTL7A), and related gene, ACTL7B, are intronless, and are located approximately 4 kb apart in a head-to-head orientation within the familial dysautonomia candidate region on 9q31. Based on mutational analysis of the ACTL7A gene in patients with this disorder, it was concluded that it is unlikely to be involved in the pathogenesis of dysautonomia. The ACTL7A gene is expressed in a wide variety of adult tissues, however, its exact function is not known. [provided by RefSeq]

Sequence: MWAPPAAIMGDGPTKKVGNQAPLQTQALQTASLRDGPAKRAVWVRHTSSEPQEPTESKAAKERPKQEVTKAVVVDLGTGYCKCGFAGLPRPTHKISTTVGKPYMETAKTGDNRKETFVGQELNNTNVHLKLVNPLRHGIIVDWDTVQDIWEYLFRQEMKIAPEEHAVLVSDPPLSPHTNREKYAEMLFEAFNTPAMHIAYQSRLSMYSYGRTSGLVVEVGHGVSYVVPIYEGYPLPSITGRLDYAGSDLTAYLLGLLNSAGNEFTQDQMGIVEDIKKKCCFVALDPIEEKKVPLSEHTIRYVLPDGKEIQLCQERFLCSEMFFKPSLIKSMQLGLHTQTVSCLNKCDIALKRDLMGNILLCGGSTMLSGFPNRLQKELSSMCPNDTPQVNVLPERDSAVWTGGSILASLQGFQPLWVHRFEYEEHGPFFLYRRCF
Especificaciones

Especificaciones

NP_006678.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
75kDa
Glutathione Sepharose 4 Fast Flow
25 ug
RUO
ACTL7A
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
10881
ACTL7A (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ACTL7A
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales