missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ACAD9 Full-length ORF (NP_054768.2, 1 a.a. - 621 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00028976-P01.10ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16168902

  • 335.00€ / 10 microgramos
Fecha estimada de envío: 11-06-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

Mitochondrial fatty acid beta-oxidation is one of the main energy-producing metabolic pathways in eukaryotes. Acyl-CoA dehydrogenases (ACADs; EC 1.3.99.13) are mitochondrial enzymes that catalyze the initial rate-limiting step in the beta-oxidation of fatty acyl-CoA. ACAD9 belongs to a group of ACADs that act on fatty acids containing 14 to 20 carbons (Zhang et al., 2002 [PubMed 12359260]).[supplied by OMIM]

Sequence: MSGCGLFLRTTAAARACRGLVVSTANRRLLRTSPPVRAFAKELFLGKIKKKEVFPFPEVSQDELNEINQFLGPVEKFFTEEVDSRKIDQEGKIPDETLEKLKSLGLFGLQVPEEYGGLGFSNTMYSRLGEIISMDGSITVTLAAHQAIGLKGIILAGTEEQKAKYLPKLASGEHIAAFCLTEPASGSDAASIRSRATLSEDKKHYILNGSKVWITNGGLANIFTVFAKTEVVDSDGSVKDKITAFIVERDFGGVTNGKPEDKLGIRGSNTCEVHFENTKIPVENILGEVGDGFKVAMNILNSGRFSMGSVVAGLLKRLIEMTAEYACTRKQFNKRLSEFGLIQEKFALMAQKAYVMESMTYLTAGMLDQPGFPDCSIEAAMVKVFSSEAAWQCVSEALQILGGLGYTRDYPYERILRDTRILLIFEGTNEILRMYIALTGLQHAGRILTTRIHELKQAKVSTVMDTVGRRLRDSLGRTVDLGLTGNHGVVHPSLADSANKFEENTYCFGRTVETLLLRFGKTIMEEQLVLKRVANILINLYGMTAVLSRASRSIRIGLRNHDHEVLLANTFCVEAYLQNLFSLSQLDKYAPENLDEQIKKVSQQILEKRAYICAHPLDRTC
Especificaciones

Especificaciones

NP_054768.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
95.2kDa
Glutathione Sepharose 4 Fast Flow
10 ug
RUO
ACAD9
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, Protein Array, ELISA, Western Blot
28976
ACAD9 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ACAD-9/FLJ23533/MGC14452/NPD002
ACAD9
Recombinant
wheat germ expression system
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales