missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human ABCA8 Full-length ORF (AAH47765.1, 1 a.a. - 266 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00010351-P01.10ug
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
The membrane-associated protein encoded by this gene is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intracellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). This protein is a member of the ABC1 subfamily. Members of the ABC1 subfamily comprise the only major ABC subfamily found exclusively in multicellular eukaryotes. The function of this protein has not yet been determined. [provided by RefSeq]
Sequence: MRKRKISVCQQTWALLCKNFLKKWRMKRESLMEWLNSLLLLLCLYIYPHSHQVNDFSSLLTMDLGRVDTFNESRFSVVYTPVTNTTQQIMNKVASTPFLAGKEVLGLPDEESIKEFTANYPEEIVRVTFTNTYSYHLKFLLGHGMPAKKEHKDHTAHCYETNEDVYCEVSVFWKEGFVALQAAINAAIIEITTNHSVMEELMSVTGKNMKMHSFIGQSGVITDLYLFSCIISFSSFIYYASVNVTRERKRMKALMTMMGLRDSAFWEspecificaciones
AAH47765.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
57.2kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
RUO | |
ABCA8 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
10351 | |
ABCA8 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
KIAA0822/MGC163152 | |
ABCA8 | |
Recombinant | |
wheat germ expression system |