missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSP20 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 16813-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
HSPB6 is associated with actin and modulates smooth muscle relaxation.Especificaciones
HSP20 | |
Polyclonal | |
Unconjugated | |
HSPB6 | |
Heat shock protein beta 6, Hsp20, HSPB6 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
126393, 192245, 243912 | |
-20°C | |
Liquid |
Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O14558, P97541, Q5EBG6 | |
Hspb6 | |
HSP20 Fusion Protein Ag10352 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |