missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HRASLS5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 17581-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
catalyzes N-acylation of phosphatidylethanolamine (PE) to generate N-Acylphosphatidylethanolamine (NAPE) a precursor of bioactive N-acylethanolamines, including the endocannabinoid adamide.Especificaciones
HRASLS5 | |
Polyclonal | |
Unconjugated | |
HRASLS5 | |
H rev107 like protein 5, HRAS like suppressor 5, HRASLS5, HRLP5, iNAT, RLP1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
117245, 293711, 66727 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q96KN8, Q9CPX5 | |
Hrasls5 | |
HRASLS5 Fusion Protein Ag11113 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |