missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HPS6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 11371-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
This intronless gene encodes a protein that may play a role in organelle biogenesis associated with melanosomes, platelet dense granules, and lysosomes. This protein interacts with Hermansky-Pudlak syndrome 5 protein. Mutations in this gene are associated with Hermansky-Pudlak syndrome type 6.Especificaciones
HPS6 | |
Polyclonal | |
Unconjugated | |
HPS6 | |
Hermansky Pudlak syndrome 6, HPS6, RP11 302K17.1, Ru, Ruby eye protein homolog | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
20170, 309446, 79803 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q7M733, Q86YV9, Q8BLY7 | |
HPS6 | |
HPS6 Fusion Protein Ag1929 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |