missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ HMGCL (Human) Recombinant Protein
Human HMGCL full-length ORF ( AAH10570, 1 a.a. - 325 a.a.) recombinant protein with GST-tag at N-terminal.
Marca: Abnova™ H00003155-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
- Sequence: MAAMRKALPRRLVGLASLRAVSTSSMGTLPKRVKIVEVGPRDGLQNEKNIVSTPVKIKLIDMLSEAGLSVIETTSFVSPKWVPQMGDHTEVLKGIQKFPGINYPVLTPNLKGFEAAVAAGAKEVVIFGAASELFTKKNINCSIEESFQRFDAILKAAQSANISVRGYVSCALGCPYEGKISPAKVAEVTKKFYSMGCYEISLGDTIGVGTPGIMKDMLSAVMQEVPLAALAVHCHDTYGQALANTLMALQMGVSVVDSSVAGLGGCPYAQGASGNLATEDLVYMLEGLGIHTGVNLQKLLEAGNFICQALNRKTSSKVAQATCKL
Especificaciones
AAH10570 | |
3-hydroxymethyl-3-methylglutaryl-Coenzyme A lyase | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HMGCL | |
GST |
3155 | |
Wheat germ expression system | |
25 μg | |
HL | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |