missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLTF Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 14286-1-AP-150UL
Informations supplémentaires : Peso : 0.00010kg
Description
This gene encodes a member of the SWI/SNF family. Members of this family have helicase and ATPase activities and are thought to regulate transcription of certain genes by altering the chromatin structure around those genes. The encoded protein contains a RING finger DNA binding motif. Two transcript variants encoding the same protein have been found for this gene. However, use of an alternative translation start site produces an isoform that is truncated at the N-terminus compared to the full-length protein.Spécification
HLTF | |
Polyclonal | |
Unconjugated | |
HLTF | |
HIP116, HIP116A, HLTF, HLTF1, RING finger protein 80, RNF80, SMARCA3, SNF2L3, ZBU1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
20585, 295568, 6596 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q14527, Q6PCN7 | |
HLTF | |
HLTF Fusion Protein Ag5596 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |