missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Euro Protection™ HI-WAY 4-in-1, High Visibility Parka
Euro Protection™ HI-WAY 4-in-1, High Visibility Parka will keep you warm, dry, and highly visible, and features a removable jacket that can be transformed into a vest.
135.00€
Especificaciones
Material | Poliéster, poliuretano |
---|---|
Color | Amarillo, azul marino |
Tipo de tapón | Cremallera |
Estructura | Costuras reforzadas |
Recubrimiento | Poliuretano Oxford 300D |
Código de producto | Marca | Tamaño | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Tamaño | Precio | Cantidad y disponibilidad | |||||
12034265
|
Euro Protection™
70549 |
Pequeño |
135.00€
cada uno |
Inicie sesión para ver el stock |
12054265
|
Euro Protection™
70551 |
Grande |
135.00€
cada uno |
Inicie sesión para ver el stock |
Descripción
- Thermal protection and waterproofness
- Fleece lining, kidney protection, central zip with flap
- Comfort (breathability, ¾ cut)
- Flexibility (compatibility with HI-WAY Jacket (70520/70530, wide range of colors)
- Parka and removable jacket that can be transformed into a vest by removing the sleeves
- Jacket is attached to the parka with the central zip and snap tabs in the collar and cuffs
- High collar
- Fixed fluorescent two-piece hood concealed in the collar and adjustable with a drawstring
- Central double slider zip, under flap closed by snaps, to the top of the collar.
- Four outside pockets: two hip pockets piped with flap closed by velcro, a vertical breast pocket, and a wallet pocket hidden under the central flap, both zipped under flap
- Inside breast patched pocket, closed with velcro
- Suspension band at the inner base of the collar
- Bands of 3M Scotchlite™ reflective material
Especificaciones
Poliéster, poliuretano | |
Cremallera | |
Poliuretano Oxford 300D | |
Resistente al agua. | |
5 (4 exteriores, 1 interior) |
Amarillo, azul marino | |
Costuras reforzadas | |
Cuello alto | |
Forro polar | |
Parka, chaqueta, chaleco |