missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HERPUD2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 20640-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
Could be involved in the unfolded protein response (UPR) pathway.Especificaciones
HERPUD2 | |
Polyclonal | |
Unconjugated | |
HERPUD2 | |
HERPUD family member 2, HERPUD2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
64224 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Immunofluorescence | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9BSE4 | |
HERPUD2 | |
HERPUD2 Fusion Protein Ag14702 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |