Learn More
Heparina, Fisher BioReagents
39.39€ - 224.07€
CAS | 9041-08-1 |
---|---|
Número MDL | MFCD00081689 |
Código de producto | Marca | Cantidad | Envase | Precio | Cantidad y disponibilidad | ||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Cantidad | Envase | Precio | Cantidad y disponibilidad | ||||
10001413
|
Fisher BioReagents
BP2524-10 |
10 mg | Poly Vial | ||||||
10429693
|
Fisher BioReagents
BP2524-50 |
50 mg | Poly Vial | ||||||
10639163
|
Fisher BioReagents
BP2524-100 |
100 mg | Vial de polietileno | ||||||
11301146
|
Fisher BioReagents
BP2524-250 |
250 mg | Poly Vial | ||||||
Descripción
La heparina se utiliza como anticoagulante en la investigación bioquímica.Identificadores de productos químicos
9041-08-1 | |
MFCD00081689 |
Especificaciones
Heparin | |
9041-08-1 | |
5.0 to 7.5 | |
MFCD00081689 | |
≈5000 | |
Powder or Solid |
From Porcine Intestinal Mucosa | |
White | |
≤5.0% | |
15, 4688 | |
Negligible |
Seguridad y manipulación
CAUTION!
Emergency Overview
Handle in accordance with good industrial hygiene and safety practice. The toxicological properties have not been fully investigated. Use personal protective equipment. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .
NFPA
Health:1
Flammability:0
Instability:0
Recommended Storage : RT