Learn More
Abnova™ HADHSC (Human) Recombinant Protein
Human HADHSC full-length ORF ( AAH00306.1, 1 a.a. - 314 a.a.) recombinant protein with GST-tag at N-terminal.
Marca: Abnova™ H00003033-P01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
- Sequence: MAFVTRQFMRSVSSSSTASASAKKIIVKHVTVIGGGLMGAGIAQVAAATGHTVVLVDQTEDILAKSKKGIEESLRKVAKKKFAENPKAGDEFVEKTLSTIATSTDAASVVHSTDLVVEAIVENLKVKNELFKRLDKFAAEHTIFASNTSSLQITSIANATTRQDRFAGLHFFNPVPVMKLVEVIKTPMTSQKTFESLVDFSKALGKHPVSCKDTPGFIVNRLLVPYLMEAIRLYERGDASKEDIDTAMKLGAGYPMGPFELLDYVGLDTTKFIVDGWHEMDAENPLHQPSPSLNKLVAENKFGKKTGEGFYKYK
Especificaciones
AAH00306.1 | |
hydroxyacyl-Coenzyme A dehydrogenase | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HADH | |
GST |
3033 | |
Wheat germ expression system | |
10 μg | |
HAD, HADH1, HADHSC, HHF4, M/SCHAD, MGC8392, SCHAD | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |