missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ HA (B/Florida/4/2006) (ACA33493, 17 a.a. - 546 a.a.) Partial Recombinant Protein with His tag expressed in 293 cells
Used for SDS-PAGE
Marca: Abnova™ P5073.50ug
Detalles adicionales : Peso : 0.02000kg
Especificaciones
ACA33493 | |
SDS-PAGE | |
HA (B/Florida/4/2006) Recombinant Protein | |
SDS-PAGE Result | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
HA | |
Recombinant | |
Mammalian cell (293) expression system | |
≥95% by SDS-PAGE |
1 mg/mL | |
Liquid | |
Mammalian cell (293) expression system | |
50 ug | |
RUO | |
Human | |
His | |
Liquid |