missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GSR Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 18257-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
This antibody is specific to GSR.
This gene encodes a member of the class-I pyridine nucleotide-disulfide oxidoreductase family. This enzyme is a homodimeric flavoprotein. It is a central enzyme of cellular antioxidant defense, and reduces oxidized glutathione disulfide (GSSG) to the sulfhydryl form GSH, which is an important cellular antioxidant. Rare mutations in this gene result in hereditary glutathione reductase deficiency. Multiple alternatively spliced transcript variants encoding different isoforms have been found.Especificaciones
GSR | |
Polyclonal | |
Unconjugated | |
GSR | |
GLUR, glutathione reductase, GRase, GRD1, GSR | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
116686, 14782, 2936 | |
-20°C | |
Liquid |
Immunofluorescence, Western Blot, Immunocytochemistry, Immunoprecipitation | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P00390, P47791, P70619 | |
Gsr | |
GSR Fusion Protein Ag13080 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |