missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPRASP2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 12159-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
The protein encoded by this gene is a member of a family that regulates the activity of G protein-coupled receptors (GPCRs). The encoded protein has been shown to be capable of interacting with several GPCRs, including the M1 muscarinic acetylcholine receptor and the calcitonin receptor. Several transcript variants encoding the same protein have been found for this gene.Especificaciones
GPRASP2 | |
Polyclonal | |
Unconjugated | |
GPRASP2 | |
GASP 2, GASP2, GPRASP2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
114928, 245607, 317405 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.48 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8BUY8, Q96D09 | |
Gprasp2 | |
GPRASP2 Fusion Protein Ag2800 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |