missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ GPLD1 (Human) Recombinant Protein
Human GPLD1 full-length ORF ( AAH20748, 1 a.a. - 176 a.a.) recombinant protein with GST-tag at N-terminal.
Marca: Abnova™ H00002822-P01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
- Sequence: MSAFRLWPGLLIMLGSLCHRGSPCGLSTHIEIGHRALEFLQLHNGRVNYRELLLEHQDAYQAGIVFPDCFYPSICKGGKFHDVSESTHWTPFLNASVHYIRENYPLPWEKDTEKLVAFLFGITSHMAADVSWHSLGLEQGFLRTMGAIDFHGSYSEAHSAGDFGTVYLHLLNFLVV
Especificaciones
AAH20748 | |
glycosylphosphatidylinositol specific phospholipase D1 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
GPLD1 | |
GST |
2822 | |
Wheat germ expression system | |
10 μg | |
GPIPLD, GPIPLDM, MGC22590, PIGPLD, PIGPLD1 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |