missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ GNRH1 (Human) Recombinant Protein
Human GNRH1 full-length ORF ( NP_000816.1, 1 a.a. - 92 a.a.) recombinant protein with GST-tag at N-terminal.
Marca: Abnova™ H00002796-P01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
- Sequence: MKPIQKLLAGLILLTWCVEGCSSQHWSYGLRPGGKRDAENLIDSFQEIVKEVGQLAETQRFECTTHQPRSPLRDLKGALESLIEEETGQKKI
Especificaciones
NP_000816.1 | |
gonadotropin-releasing hormone 1 (luteinizing-releasing hormone) | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
GNRH1 | |
GST |
2796 | |
Wheat germ expression system | |
10 μg | |
GNRH, GRH, LHRH, LNRH | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |