missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ GCK (Human) Recombinant Protein
Human GCK full-length ORF ( AAH01890, 1 a.a. - 465 a.a.) recombinant protein with GST-tag at N-terminal.
Marca: Abnova™ H00002645-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
- Sequence: MLDDRARMEAAKKEKVEQILAEFQLQEEDLKKVMRRMQKEMDRGLRLETHEEASVKMLPTYVRSTPEGSEVGDFLSLDLGGTNFRVMLVKVGEGEEGQWSVKTKHQMYSIPEDAMTGTAEMLFDYISECISDFLDKHQMKHKKLPLGFTFSFPVRHEDIDKGILLNWTKGFKASGAEGNNVVGLLRDAIKRRGDFEMDVVAMVNDTVATMISCYYEDHQCEVGMIVGTGCNACYMEEMQNVELVEGDEGRMCVNTEWGAFGDSGELDEFLLEYDRLVDESSANPGQQLYEKLIGGKYMGELVRLVLLRLVDENLLFHGEASEQLRTRGAFETRFVSQVESDTGDRKQIYNILSTLGLRPSTTDCDIVRRACESVSTRAAHMCSAGLAGVINRMRESRSEDVMRITVGVDGSVYKLHPSFKERFHASVRRLTPSCEITFIESEEGSGRGAALVSAVACKKACMLGQ
Especificaciones
AAH01890 | |
glucokinase (hexokinase 4) | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
GCK | |
GST |
2645 | |
Wheat germ expression system | |
25 μg | |
GK, GLK, HHF3, HK4, HKIV, HXKP, MODY2 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |