missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GCHFR Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 18809-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
Mediates tetrahydrobiopterin inhibition of GTP cyclohydrolase 1. This inhibition is reversed by L-phenylalanine.Especificaciones
GCHFR | |
Polyclonal | |
Unconjugated | |
GCHFR | |
GCHFR, GFRP, HsT16933, P35 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
171128, 2644, 320415 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.22 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P30047, P70552, P99025 | |
Gchfr | |
Peptide | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |