missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GAS41 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 11050-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
The protein encoded by this gene is found in the nucleoli. It has high sequence homology to human MLLT1, and yeast and human MLLT3 proteins. Both MLLT1 and MLLT3 proteins belong to a class of transcription factors, indicating that the encoded protein might also represent a transcription factor. This protein is thought to be required for RNA transcription. This gene has been shown to be amplified in tumors. Alternative splicing results in multiple transcript variants encoding different isoforms.Especificaciones
GAS41 | |
Polyclonal | |
Unconjugated | |
YEATS4 | |
4930573H17Rik, B230215M10Rik, GAS41, Glioma amplified sequence 41, NUBI 1, NuBI1, NuMA binding protein 1, YAF9, YEATS domain containing 4, YEATS4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
299810, 64050, 8089 | |
-20°C | |
Liquid |
Western Blot | |
0.37 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O95619, Q9CR11 | |
Yeats4 | |
GAS41 Fusion Protein Ag1524 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |