Learn More
gamma tubulin Rabbit anti-Canine, Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 26195-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
This antibody can label the centrosome structure.
This gene encodes a protein that interacts with thyroid hormone receptors, also known as hormone-dependent transcription factors. The gene product interacts specifically with the ligand binding domain. This gene is one of several that may play a role in early-stage non-small cell lung cancer.Especificaciones
gamma tubulin | |
Polyclonal | |
Unconjugated | |
TUBG1 | |
Gamma 1 tubulin, GCP 1, TUBG, TUBG1, TUBGCP1, Tubulin gamma 1 chain, tubulin, gamma 1, tubulin-gamma | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
103733, 403695, 7283 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.29 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P23258, P83887, Q9GKK5 | |
TUBG1 | |
gamma tubulin Fusion Protein Ag23575 | |
150 μL | |
Primary | |
Canine, Human, Mouse | |
Antibody | |
IgG |