missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GAL3ST1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 18503-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
Catalyzes the sulfation of membrane glycolipids. Seems to prefer beta-glycosides at the non-reducing termini of sugar chains attached to a lipid moiety. Catalyzes the synthesis of galactosylceramide sulfate (sulfatide), a major lipid component of the myelin sheath and of monogalactosylalkylacylglycerol sulfate (seminolipid), present in spermatocytes. Also acts on lactosylceramide, galactosyl 1-alkyl-2-sn-glycerol and galactosyl diacylglycerol (in vitro).Especificaciones
GAL3ST1 | |
Polyclonal | |
Unconjugated | |
GAL3ST1 | |
Cerebroside sulfotransferase, CST, GAL3ST1, GalCer sulfotransferase | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
53897, 683713, 9514 | |
-20°C | |
Liquid |
Western Blot | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q99999, Q9JHE4 | |
Gal3st1 | |
GAL3ST1 Fusion Protein Ag13353 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |