missing translation for 'onlineSavingsMsg'
Learn More
Learn More
G3BP2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 16276-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
G3BP2 gene ontology annotations related to this gene include stress granule assembly.Especificaciones
G3BP2 | |
Polyclonal | |
Unconjugated | |
G3BP2 | |
G3BP 2, G3BP2, KIAA0660 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
9908 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9UN86 | |
G3BP2 | |
G3BP2 Fusion Protein Ag9355 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |