missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FXYD3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 15853-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
FXYD3 belongs to a small family of FXYD-domain containing regulators of Na+/K+ ATPases which share a 35-amino acid signature sequence domain, beginning with the sequence PFXYD, and containing 7 invariant and 6 highly conserved amino acids. This gene encodes a cell membrane protein that may regulate the function of ion-pumps and ion-channels. This gene may also play a role in tumor progression. Alternative splicing results in multiple transcript variants encoding distinct isoforms.Especificaciones
FXYD3 | |
Polyclonal | |
Unconjugated | |
FXYD3 | |
FXYD3, Mammary tumor 8 kDa protein, MAT 8, MAT8, Phospholemman like, PLML | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
116831, 17178, 5349 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.19 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P59645, Q14802, Q61835 | |
Fxyd3 | |
FXYD3 Fusion Protein Ag8672 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |