missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FRK (PTK5) Recombinant Human Protein
FRK (PTK5) Recombinant Human Protein
Marca: Invitrogen PV3874
Detalles adicionales : Peso : 0.05250kg
Descripción
FRK (PTK5) is a nuclear protein and may function during G1 and S phase of the cell cycle, suppressing growth.Especificaciones
NP_002022 | |
Pharma and Biopharma, Protein Assays and Analysis, Target and Lead Identification and Validation, Kinase Assay | |
86.3kDa | |
10μg | |
For research use only. Not for use in diagnostic procedures | |
FRK (PTK5), GTK, RAK | |
FRK (PTK5) was pre-diluted in enzyme dilution buffer and assayed in 25mM Tris (pH 7.5), 10mM MgCl2, 0.5mM EGTA, 0.5mM Na3VO4, 5mM β-glycerophosphate, 2.5mM DTT, 0.01% Triton™ X-100, 200μM ATP, 200μg/mL poly [Glu, Tyr] 4:1 substrate and trace [32P]-γ-ATP for 10 minutes at 30°C | |
FRK (PTK5) Recombinant Human Protein | |
Human | |
Baculovirus | |
Cytoplasmic tyrosine kinase |
0.22mg/mL | |
114294, 2444 | |
FRK (PTK5) Recombinant Human Protein | |
–80°C | |
1 Each | |
FRK (PTK5) Recombinant Human Protein | |
SRC | |
100nmoles of phosphate transferred to poly [Glu, Tyr] 4:1 substrate per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 4μg/mL | |
GST Tag | |
Kinase and inhibitors | |
75% by SDS-PAGE |
For Research Use Only. Not for use in diagnostic procedures.