missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Folate Receptor alpha Monoclonal Antibody (01), Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA543018
Detalles adicionales : Peso : 0.01000kg
Descripción
Specificity: Human FOLR1
Folate Receptor alpha Monoclonal antibody specifically detects Folate Receptor alpha in Human samples. It is validated for ELISA.Especificaciones
Folate Receptor alpha | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
P15328 | |
FOLR1 | |
Recombinant Human FOLR1 Protein. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA | |
01 | |
Unconjugated | |
FOLR1 | |
66/67kDa; AA407891; adult folate-binding protein; eIF3 p66; EIF3D; eIF3d {ECO:0000255; eIF3p66; eIF3-p66; EIF3S7; eIF-3-zeta; eIF3-zeta; eIF-3-zeta {ECO:0000255; Eukaryotic translation initiation factor 3 subunit 7; eukaryotic translation initiation factor 3 subunit 7 {ECO:0000255; eukaryotic translation initiation factor 3 subunit D; eukaryotic translation initiation factor 3 subunit D {ECO:0000255; eukaryotic translation initiation factor 3, subunit 7 (zeta); eukaryotic translation initiation factor 3, subunit 7 zeta, 66/67kDa; eukaryotic translation initiation factor 3, subunit D; FBP; Fbp1; folate binding protein; folate binding protein 1; folate receptor 1; folate receptor 1 (adult); folate receptor alpha; Folate receptor, adult; folate-binding protein 1; Folbp1; Folbp-1; FOLR; FOLR1; FR-alpha; HAMAP-Rule:MF_03003}; KB cells FBP; MOv18; Ovarian tumor-associated antigen MOv18; translation initiation factor eIF3 p66; translation initiation factor eIF3 p66 subunit | |
Mouse | |
Protein A | |
RUO | |
2348 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |