missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ FKBP5 (Human) Recombinant Protein
Human FKBP5 full-length ORF ( AAH42605, 1 a.a. - 457 a.a.) recombinant protein with GST-tag at N-terminal.
Marca: Abnova™ H00002289-P01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
- Sequence: MTTDEGAKNNEESPTATVAEQGEDITSKKDRGVLKIVKRVGNGEETPMIGDKVYVHYKGKLSNGKKFDSSHDRNEPFVFSLGKGQVIKAWDIGVATMKKGEICHLLCKPEYAYGSAGSLPKIPSNATLFFEIELLDFKGEDLFEDGGIIRRTKRKGEGYSNPNEGATVEIHLEGRCGGRMFDCRDVAFTVGEGEDHDIPIGIDKALEKMQREEQCILYLGPRYGFGEAGKPKFGIEPNAELIYEVTLKSFEKAKESWEMDTKEKLEQAAIVKEKGTVYFKGGKYMQAVIQYGKIVSWLEMEYGLSEKESKASESFLLAAFLNLAMCYLKLREYTKAVECCDKALGLDSANEKGLYRRGEAQLLMNEFESAKGDFEKVLEVNPQNKAARLQISMCQKKAKEHNERDRRIYANMFKKFAEQDAKEEANKAMGKKTSEGVTNEKGTDSQAMEEEKPEGHV
Especificaciones
AAH42605 | |
FK506 binding protein 5 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FKBP5 | |
GST |
2289 | |
Wheat germ expression system | |
10 μg | |
FKBP51, FKBP54, MGC111006, P54, PPIase, Ptg-10 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |