missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems FITC-Labeled Human ROR1 Protein, Fc Tag

Marca:  ACROBiosystems RO1-HF253-25ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 17196651

  • 363.80€ / 25 microgramos
Fecha estimada de envío: 20-10-2022
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

Tyrosine-protein kinase transmembrane receptor ROR1 is also known as Neurotrophic tyrosine kinase, receptor-related 1 (NTRKR1), which belongs to the protein kinase superfamily or tyr protein kinase family or ROR subfamily. ROR1 contains 1 FZ (frizzled) domain, 1 Ig-like C2-type (immunoglobulin-like) domain, 1 kringle domain, 1 protein kinase domain. ROR1 is expressed at high levels during early embryonic development. The expression levels drop strongly around day 16 and there are only very low levels in adult tissues. Isoform Short is strongly expressed in fetal and adult CNS and in a variety of human cancers, including those originating from CNS or PNS neuroectoderm. ROR1 could interact with casein kinase 1 epsilon (CK1ε) to activate phosphoinositide 3-kinase-mediated AKT phosphorylation and cAMP-response-element-binding protein (CREB), which was associated with enhanced tumor-cell growth.

This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 68.6 kDa. The protein migrates as 80-90 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Especificaciones

Especificaciones

Powder
>90% as determined by SDS-PAGE
FITC-Labeled Human ROR1, Fc Tag (RO1-HF253) is expressed from human 293 cells (HEK293). It contains AA Gln 30 - Glu 403 (Accession # Q01973-1). It is the FITC labeled form of Human ROR1, Fc Tag (RO1-H5250).
-20°C, avoid light
ROR1
Fc Tag
Less than 1.0 EU / µg by the LAL method.
Fluorescent labeled protein
68.6 kDa
25 µg
Gln 30 - Glu 403
ROR1,NTRKR1
FITC Labeled Protein
PBS, pH7.4
HEK293
Human
Videos
SDS
Documentos
Certificados
Ofertas especiales

Ofertas especiales