Learn More
ACROBiosystems FITC-Labeled Human GUCY2C / Guanylyl cyclase C Protein, His Tag
Marca:  ACROBiosystems GUC-HF2H8-25ug
Detalles adicionales : Peso : 0.00010kg
 Ver más versiones de este producto
Código de producto. 17186351
  • 363.80€ / 25 microgramos
Fecha estimada de envío: 19-10-2022
para ver el stock
Descripción
Descripción
GUCY2C (Guanylyl Cyclase C), also known as heat-stable enterotoxin receptor, is a type I transmembrane protein of the guanylate cyclase (gc) family that signal by producing cGMP. Guanylate cyclase C (GUCY2C) and its hormones guanylin and uroguanylin have recently emerged as one paracrine axis defending intestinal mucosal integrity against mutational, chemical, and inflammatory injury. GUCY2C murine CAR-T cells recognized and killed human colorectal cancer cells endogenously expressing GUCY2C. Thus, we have identified a human GUCY2C-specific CAR-T cell therapy approach that may be developed for the treatment of GUCY2C-expressing metastatic colorectal cancer.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 47.9 kDa. The protein migrates as 60-90 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Especificaciones
Especificaciones
NP_004954.2
47.9 kDa
25 µg
Ser 24 - Gln 430
GUCY2C,GUC2C,STAR,STA receptor,hSTAR,GC-C
FITC Labeled Protein
PBS, pH7.4
HEK293
Human
Powder
>95% as determined by SDS-PAGE
FITC-Labeled Human GUCY2C, His Tag (GUC-HF2H8) is expressed from human 293 cells (HEK293). It contains AA Ser 24 - Gln 430 (Accession # P25092-1). It is the FITC labeled form of Human GUCY2C, His Tag (GUC-H52H5).
-20°C, avoid light
GUCY2C
His Tag
Less than 1.0 EU / µg by the LAL method.
Fluorescent labeled protein