Learn More
ACROBiosystems FITC-Labeled Human Glypican 3 / GPC3 Protein, Fc Tag
Marca: ACROBiosystems GP3-HF258-25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Glypican-3 (GPC3) is also known as Intestinal protein OCI-5, GTR2-2, MXR7, which belongs to the glypican family. Glypican 3 / GPC-3 is highly expressed in lung, liver and kidney. Glypican-3 inhibits the dipeptidyl peptidase activity of DPP4. Glypican-3 may be involved in the suppression/modulation of growth in the predominantly mesodermal tissues and organs, and also may play a role in the modulation of IGF2 interactions with its receptor and thereby modulate its function.
This protein carries a human IgG1 Fc tag at the C-terminus. This protein contains a furin-like convertase cleavage site, 355-RQYR-358, and will be partially processed into N and C-terminal fragment with calculated MW of 38.1 kDa and 49.3 kDa respectivelyEspecificaciones
NP_004475.1 | |
25 μg | |
Gln 25 - His 559 | |
GPC3,OCI5,Glypican-3,GTR2-2,MXR7,DGSX,SDYS ,SGB,SGBS,SGBS1 | |
FITC | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |
38.1 kDa and 49.3 kDa | |
FITC-Labeled Human Glypican 3, Fc Tag (Cat. No. GP3-HF258) is expressed from human HEK293 cells. It contains AA Gln 25 - His 559 (Accession # P51654-1). It is the FITC labeled form of Human Glypican 3, Fc Tag (Cat. No. GP3-H5258). | |
-20°C, avoid light | |
Glypican 3 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Fluorescent labeled protein |