Learn More
ACROBiosystems FITC-Labeled Human CD4 Protein, Fc Tag
Marca: ACROBiosystems CD4-HF255-25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
T-cell surface glycoprotein CD4 is also known as T-cell surface antigen T4/Leu-3. CD4 contains three Ig-like C2-type (immunoglobulin-like) domains and one Ig-like V-type (immunoglobulin-like) domain. CD4 is accessory protein for MHC class-II antigen/T-cell receptor interaction. CD4 induces the aggregation of lipid rafts. CD4 is a primary receptor used by HIV-1 to gain entry into host T cells. HIV infection leads to a progressive reduction of the number of T cells possessing CD4 receptors. Therefore, medical professionals refer to the CD4 count to decide when to begin treatment for HIV-infected patients.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 67.7 kDa. The protein migrates as 67-80 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Especificaciones
AAH25782 | |
25 μg | |
Lys 26 - Pro 396 | |
CD4,CD4mut,LEU3 | |
FITC | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
67.7 kDa | |
FITC-Labeled Human CD4, Fc Tag (CD4-HF255) is expressed from human 293 cells (HEK293). It contains AA Lys 26 - Pro 396 (Accession # P01730). It is the FITC labeled form of Human CD4, Fc Tag (CD4-H5259). | |
-20°C, avoid light | |
CD4 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Fluorescent labeled protein |