missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ferritin, Light Chain Mouse, Clone: FTL/1389, enQuireBio™
Mouse Monoclonal Antibody
Marca: EnquireBio 2512-MSM4-P1
Detalles adicionales : Peso : 0.01000kg
Especificaciones
Ferritin, Light Chain | |
Cytoplasmic | |
FTL/1389 | |
Unconjugated | |
NP_000137 | |
Mouse | |
19-25kDa | |
Protein A/G Purified | |
RUO | |
2512 | |
IgG1 |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
FTL | |
Recombinant human FTL protein fragment (aa 38-165) | |
HepG2, HeLa, HL-60 or 293T cells. Pancreas, Liver, Cerebellum or Testis. | |
100 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |