Learn More
FCER1A Armenian Hamster, PE, Clone: 42795, enQuireBio™
Armenian Hamster Monoclonal Antibody
Marca: EnquireBio QAB91-PE-25ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
FCER1A | |
Monoclonal | |
0.5mg/mL | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
FCER1A | |
Protein A/G Purified | |
RUO | |
2205 | |
IgG |
Flow Cytometry | |
42795 | |
PE | |
NP_001992 | |
Armenian Hamster | |
25 μg | |
Primary | |
Store at 2-4°C. |