missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM131B Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 26498-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
FAM131B is a protein coding gene.Especificaciones
FAM131B | |
Polyclonal | |
Unconjugated | |
FAM131B | |
FAM131B, KIAA0773, Protein FAM131B | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
9715 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q86XD5 | |
FAM131B | |
FAM131B Fusion Protein Ag24143 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |