missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Exo70 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 12014-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
The protein encoded by this gene is a component of the exocyst complex. The exocyst complex plays a critical role in vesicular trafficking and the secretory pathway by targeting post-Golgi vesicles to the plasma membrane. The encoded protein is required for assembly of the exocyst complex and docking of the complex to the plasma membrane. The encoded protein may also play a role in pre-mRNA splicing through interactions with pre-mRNA-processing factor 19. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene, and a pseudogene of this gene is located on the long arm of chromosome 4.Especificaciones
Exo70 | |
Polyclonal | |
Unconjugated | |
EXOC7 | |
2 5 3p, EX070, Exo70, Exo70p, EXOC1, EXOC7, exocyst complex component 7, KIAA1067, YJL085W | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23265, 53413, 64632 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O35250, O54922, Q9UPT5 | |
Exoc7 | |
Exo70 Fusion Protein Ag2628 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |