missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EXD1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 17313-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
This antibody can recognize all these three isoforms.
EXD1 is an RNA-binding component of the PET complex, a multiprotein complex required for the processing of piRNAs during spermatogenesis. The piRNA metabolic process mediates the repression of transposable elements during meiosis by forming complexes composed of piRNAs and Piwi proteins and governs the methylation and subsequent repression of transposable elements, preventing their mobilization, which is essential for the germline integrity. The PET complex is required during the secondary piRNAs metabolic process for the PIWIL2 slicing-triggered loading of PIWIL4 piRNAs. In the PET complex, EXD1 probably acts as an RNA adapter. EXD1 is an inactive exonuclease.Especificaciones
EXD1 | |
Polyclonal | |
Unconjugated | |
EXD1 | |
EXD1, EXDL1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
161829, 241624, 311334 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8CDF7, Q8NHP7 | |
Exd1 | |
EXD1 Fusion Protein Ag11282 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |