missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EVI2A Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 17415-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descrizione
May complex with itself or/and other proteins within the membrane, to function as part of a cell-surface receptor.Specifica
EVI2A | |
Polyclonal | |
Unconjugated | |
EVI2A | |
EVDA, EVI 2A, EVI2, EVI2A, Protein EVI2A | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
2123 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.15 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P22794 | |
EVI2A | |
EVI2A Fusion Protein Ag11075 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |