missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ ETV4 (Human) Recombinant Protein
Human ETV4 full-length ORF ( AAH07242, 1 a.a. - 207 a.a.) recombinant protein with GST-tag at N-terminal.
Marca: Abnova™ H00002118-P01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
- Sequence: MYLHTEGFSGPSPGDGAMGYGYEKPLRPFPDDVCVAPEKFEGDIKQEGVGAFREGPPYQRRGALQLWQFLVALLDDPTNAHFIAWTGRGMEFKLIEPEEVARLWGIQKNRPAMNYDKLSRSLRYYYEKGIMQKVAGERYVYKFVCEPEALFSLAFPDNQRPALKAEFDRPVSEEDTVPLSHLDESPAYLPELAGPAQPFGPKGGYSY
Especificaciones
AAH07242 | |
ets variant 4 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ETV4 | |
GST |
2118 | |
Wheat germ expression system | |
10 μg | |
E1A-F, E1AF, PEA3, PEAS3 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |