missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ETS1 Mouse anti-Human, Rat, Clone: 4F2D4, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 66598-1-IG-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
Principal Names: c-Ets-1; v-ets erythroblastosis virus E26 oncogene homlog 1 (avian); ETS-1; EWSR2; p54; Official Gene Symbol: ETS1 Gene ID: 2113 (Human) Gene Map Locus: 11q23.3 (Human) ETS1, a 441 A.A protein, is a member of DNA-binding ETS protein family. It is a multifunctional protein consisting of an N-terminal pointed domain, a central transactivation domain and 2 auto inhibitory domains flanked on both sides of conserved C-terminal Ets domain. It interacts with factors including PEBP2alphaA, p300/CBP and mediates transactivation of Osteopontin, Presenilin-1, Stromelysin and Collagenase. It is activated by Ras/MAPK signaling and its expression is up-regulated in resting T-cells. Its activity is modulated by various factors including AP-1, Sp-1, c-myb and MafB. Sp100 activates ETS1 in nuclear bodies. It interacts with EAP1/Daxx and causes repression of MMP1 and BCL2 transactivation. It interacts with STAT6 and modulates Socs-1 expression induced by IL-4. It interacts with GATA3 and positively regulates Tax-1 dependant IL-5 expression.Especificaciones
ETS1 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P14921, P41156 | |
ETS1 | |
ETS1 Fusion Protein Ag2127 | |
150 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG2b |
Western Blot | |
4F2D4 | |
Unconjugated | |
ETS1 | |
ETS 1, ETS1, EWSR2, p54, Protein C ets 1 | |
Mouse | |
Protein A | |
RUO | |
2113, 24356 | |
-20°C | |
Liquid |