missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Enkurin Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 26440-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
Enkurin interacts with transient receptor potential canonical.Especificaciones
Enkurin | |
Polyclonal | |
Unconjugated | |
ENKUR | |
C10orf63, ENKUR, Enkurin | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
219670, 71233 | |
-20°C | |
Liquid |
Western Blot | |
0.28 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q6SP97, Q8TC29 | |
Enkur | |
Enkurin Fusion Protein Ag23762 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |