missing translation for 'onlineSavingsMsg'
Learn More
Learn More
endoglin / CD105 Mouse, Clone: ENG/1327, enQuireBio™
Mouse Monoclonal Antibody
Marca: EnquireBio 2022-MSM2-P1
Detalles adicionales : Peso : 0.01000kg
Especificaciones
endoglin / CD105 | |
Cytoplasmic | |
ENG/1327 | |
Unconjugated | |
NP_001265067 | |
Mouse | |
84kDa | |
Protein A/G Purified | |
RUO | |
2022 | |
IgG1 |
Western Blot | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
ENG | |
Recombinant human endoglin protein fragment (aa74-251) | |
HeLa, A431 and HL-60 cells. Colon. | |
100 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |