missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ ECHS1 (Human) Recombinant Protein
Human ECHS1 full-length ORF ( AAH08906, 13 a.a. - 290 a.a.) recombinant protein with GST-tag at N-terminal.
Marca: Abnova™ H00001892-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
- Sequence: GPLRPPVRCPAWRPFASGANFEYIIAEKRGKNNTVGLIQLNRPKALNALCDGLIDELNQALKIFEEDPAVGAIVLTGGDKAFAAGADIKEMQNLSFQDCYSSKFLKHWDHLTQVKKPVIAAVNGYAFGGGCELAMMCDIIYAGEKAQFAQPEILIGTIPGAGGTQRLTRAVGKSLAMEMVLTGDRISAQDAKQAGLVSKICPVETLVEEAIQCAEKIASNSKIVVAMAKESVNAAFEMTLTEGSKLEKKLFYSTFATDDRKEGMTAFVEKRKANFKDQ
Especificaciones
AAH08906 | |
enoyl Coenzyme A hydratase, short chain, 1, mitochondrial | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ECHS1 | |
GST |
1892 | |
Wheat germ expression system | |
25 μg | |
SCEH | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |