missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DYDC1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26327-1-AP-150UL
Additional Details : Peso : 0.00010kg
Description
This gene encodes a member of a family of proteins that contains a DPY30 domain. The encoded protein is involved in acrosome formation during spermatid development. This gene locus overlaps with a closely related gene on the opposite strand. Alternative splicing results in multiple transcript variants.Specifications
DYDC1 | |
Polyclonal | |
Unconjugated | |
DYDC1 | |
DPY30 domain containing 1, DPY30D1, DYDC1, RSD 9 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
143241, 69496 | |
-20°C | |
Liquid |
Western Blot | |
0.53 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8WWB3, Q9D9T0 | |
Dydc1 | |
DYDC1 Fusion Protein Ag23693 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |