Learn More
Thermo Scientific™ Dog (Beagle) S9 Fractions
Liver S9 fractions prepared from a pool of male Beagle Dogs (total protein concentration: 20mg/mL).
Marca: Thermo Scientific™ DGS9PL
Detalles adicionales : Peso : 0.99000kg
Descripción
We provide liver subcellular fractions from a variety of tox species, including human, nonhuman primates (Cynomolgus Monkey and Rhesus Monkey), dog (Beagle), rat (Sprague-Dawley), mouse (CD-1 and Balb-c), and trout (Oncorhynchus mykiss). Custom species are available upon request.
Especificaciones
20 mg⁄ml | |
1 mL | |
Store between -60°C and -85°C. | |
Male | |
S9 Fraction | |
Frozen | |
Research Grade |
Tube | |
Dog/Canine | |
Pooled | |
Dry Ice | |
Liver | |
Gibco™ |