missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DOG-1 / TMEM16A Mouse, Clone: DG1/1486, enQuireBio™
Mouse Monoclonal Antibody
Marca: EnquireBio 55107-MSM6-P0
Detalles adicionales : Peso : 0.01000kg
Especificaciones
DOG-1 / TMEM16A | |
Cell Surface and Cytoplasm | |
DG1/1486 | |
Unconjugated | |
NP_011543428 | |
Mouse | |
114kDa | |
Protein A/G Purified | |
RUO | |
55107 | |
IgG2b κ |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
TMEM16A | |
Recombinant human DOG-1 protein fragment (aa 2-101) | |
Gastrointestinal Stromal Tumor (GIST) or testicular germ cell tumor. Melanocytes in the basal layer of the epidermis and mast cells in the dermis of normal skin. | |
20 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |