missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ DMWD (Human) Recombinant Protein
Human DMWD full-length ORF ( AAH19266.1, 1 a.a. - 334 a.a.) recombinant protein with GST-tag at N-terminal.
Marca: Abnova™ H00001762-P01.10ug
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
- Sequence: MLLRGLMKSYFGGLLCVCWSPDGRYVVTGGEDDLVTVWSFTEGRVVARGHGHKSWVNAVAFDPYTTRAEEAATAAGADGERSGEEEEEEPEAAGTGSAGGAPLSPLPKAGSITYRFGSAGQDTQFCLWDLTEDVLYPHPPLARTRTLPGTPGTTPPAASSSRGGEPGPGPLPRSLSRSNSLPHPAGGGKAGGPGVAAEPGTPFSIGRFATLTLQERRDRGAEKEHKRYHSLGNISRGGSGGSGSGGEKPSGPVPRSRLDPAKVLGTALCPRIHEVPLLEPLVCKKIAQERLTVLLFLEDCIITACQEGLICTWARPGKAGISSQPGNSPSGTVV
Especificaciones
AAH19266.1 | |
dystrophia myotonica, WD repeat containing | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DMWD | |
GST |
1762 | |
Wheat germ expression system | |
10 μg | |
D19S593E, DMR-N9, DMRN9, gene59 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |