missing translation for 'onlineSavingsMsg'
Learn More

DLL3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 25535-1-AP-20UL

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16824105

  • 140.00€ / 20 microlitros
Fecha estimada de envío: 15-05-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

Delta-like protein 3 is a ligand for the Notch signaling pathway. It inhibits primary neurogenesis. DLL3 plays a role in the formation of somite boundaries during segmentation of the paraxial mesoderm. Defects in DLL3 are the cause of Spondylocostal dysostosis autosomal recessive type 1 (SCDO1). Mutations in DLL3 gene cause truncal shortening relative to their limbs, which leads to abdominal protrusion, abnormal spinal curvature and sometimes a plagiocephaly-torticollis sequence. It may be required to divert neurons along a specific differentiation pathway.
Especificaciones

Especificaciones

DLL3
Polyclonal
Unconjugated
DLL3
delta like 3 (Drosophila), Delta like protein 3, Delta3, DLL3, Drosophila Delta homolog 3, SCDO1
Rabbit
Antigen Affinity Chromatography
RUO
10683, 13389
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin)
0.37 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O88516, Q9NYJ7
DLL3
DLL3 Fusion Protein Ag21965
20 μL
Primary
Human, Mouse
Antibody
IgG
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales