missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DLG5 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 15687-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
May play a role at the plasma membrane in the maintece of the structure of epithelial cells and in the transmission of extracellular signals to the membrane and cytoskeleton.Especificaciones
DLG5 | |
Polyclonal | |
Unconjugated | |
DLG5 | |
Discs large protein P dlg, Disks large homolog 5, DLG5, KIAA0583, LP DLG, P DLG5, PDLG, Placenta and prostate DLG | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
71228, 9231 | |
-20°C | |
Liquid |
Western Blot | |
0.29 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
E9Q9R9, Q8TDM6 | |
Dlg5 | |
DLG5 Fusion Protein Ag8194 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |