missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DLG1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 55085-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
The antibody is specific to DLG1.
This gene encodes a multi-domain scaffolding protein that is required for normal development. This protein may have a role in septate junction formation, signal transduction, cell proliferation, synaptogenesis and lymphocyte activation. Several alternatively spliced transcript variants encoding different isoforms have been described for this gene, but the full-length nature of some of the variants is not known.Especificaciones
DLG1 | |
Polyclonal | |
Unconjugated | |
DLG1 | |
Disks large homolog 1, DLG1, DLGH1, hdlg, SAP 97, SAP97, Synapse associated protein 97 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
13383, 1739, 25252 | |
-20°C | |
Liquid |
Western Blot | |
0.27 mg/mL | |
0.1 M glycine, 0.1 M sodium bicarbonate with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q12959, Q62696, Q811D0 | |
DLG1 | |
Peptide | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |