missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DHPS Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 11184-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes a protein that is required for the formation of hypusine, a unique amino acid formed by the posttranslational modification of only one protein, eukaryotic translation initiation factor 5A. The encoded protein catalyzes the first step in hypusine formation by transferring the butylamine moiety of spermidine to a specific lysine residue of the eukaryotic translation initiation factor 5A precursor, forming an intermediate deoxyhypusine residue. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.Especificaciones
DHPS | |
Polyclonal | |
Unconjugated | |
DHPS | |
deoxyhypusine synthase, DHPS, DHS, DS, MIG13 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
1725, 288923, 330817 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
0.29 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P49366, Q3TXU5, Q6AY53 | |
Dhps | |
DHPS Fusion Protein Ag1646 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |